CAS RN 160896-49-1
CAS RN 160896-49-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Hosenkoside K, MedChemExpress
MedChemExpress Hosenkoside K is a baccharane glycoside isolated from the seeds of impatiens balsamina.