CAS RN 1604810-84-5
CAS RN 1604810-84-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
THZ2, MedChemExpress
MedChemExpress THZ2 is a potent and selective CDK7 inhibitor with an IC50 of 13.9 nM.