accessibility menu, dialog, popup

UserName

CAS RN 15933-07-0

H3COOOCH3

CAS RN 15933-07-0

IUPAC Name: ethyl 2-oxobutanoate
Synonyms: ethyl 2-oxobutanoate
Molecular Weight (g/mol): 130.14
Molecular Formula: C6H10O3
InChi Key: FJAKCEHATXBFJT-UHFFFAOYSA-N
SMILES: CCOC(=O)C(=O)CC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results