CAS RN 1573-28-0
CAS RN 1573-28-0
IUPAC Name:
3,6-dimethylheptan-3-ol
Synonyms:
3-heptanol, 3,6-dimethyl-
Molecular Weight (g/mol):
144.26
Molecular Formula:
C9H20O
InChi Key:
HVPGGLNDHUWMLS-UHFFFAOYNA-N
SMILES:
CCC(C)(O)CCC(C)C
1
–
1
of
1
results
1
3,6-Dimethyl-3-heptanol, 99%, Thermo Scientific™
CAS: 1573-28-0 Molecular Formula: C9H20O Molecular Weight (g/mol): 144.26 MDL Number: MFCD00021828 InChI Key: HVPGGLNDHUWMLS-UHFFFAOYNA-N PubChem CID: 15297 IUPAC Name: 3,6-dimethylheptan-3-ol SMILES: CCC(C)(O)CCC(C)C