CAS RN 15694-70-9
CAS RN 15694-70-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Nickel(II) formate dihydrate
CAS: 15694-70-9 Molecular Formula: Ni(HCO2)2·2H2O MDL Number: MFCD00153034