CAS RN 156453-51-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(R)-Dapoxetine Hydrochloride, TRC
CAS: 156453-51-9 Molecular Formula: C21 H23 N O . Cl H Molecular Weight (g/mol): 341.87 Synonym: (αR)-N,N-Dimethyl-α-[2-(1-naphthalenyloxy)ethyl]-benzenemethanamine Hydrochloride IUPAC Name: (1R)-N,N-dimethyl-3-(1-naphthyloxy)-1-phenyl-propan-1-amine;hydrochloride SMILES: Cl.CN(C)[C@H](CCOc1cccc2ccccc12)c3ccccc3