accessibility menu, dialog, popup

UserName

CAS RN 1558-25-4

ClSiClClCl

CAS RN 1558-25-4

IUPAC Name: trichloro(chloromethyl)silane
Synonyms: trichloro(chloromethyl)silane
Molecular Weight (g/mol): 183.91
Molecular Formula: CH2Cl4Si
InChi Key: FYTPGBJPTDQJCG-UHFFFAOYSA-N
SMILES: ClC[Si](Cl)(Cl)Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results