CAS RN 15572-25-5
CAS RN 15572-25-5
IUPAC Name:
bis(λ¹-thallanylium) selandiide
Synonyms:
bis(λ¹-thallanylium) selandiide
Molecular Weight (g/mol):
487.73
Molecular Formula:
SeTl2
InChi Key:
KLNGSAIQZVCZLH-UHFFFAOYSA-N
SMILES:
[Se--].[Tl+].[Tl+]
1
–
1
of
1
results
1
Thallium(I) selenide, 99.9% (metals basis)
CAS: 15572-25-5 MDL Number: MFCD00049603