accessibility menu, dialog, popup

UserName

CAS RN 152305-23-2

HNOOH2N

CAS RN 152305-23-2

IUPAC Name: 4-[(4-aminophenyl)methyl]-1,3-oxazolidin-2-one
Synonyms: 4-[(4-aminophenyl)methyl]-1,3-oxazolidin-2-one
Molecular Weight (g/mol): 192.22
Molecular Formula: C10H12N2O2
InChi Key: WNAVSKJKDPLWBD-UHFFFAOYNA-N
SMILES: NC1=CC=C(CC2COC(=O)N2)C=C1
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
Industry Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results