accessibility menu, dialog, popup

UserName

CAS RN 15086-94-9

HOBrOBrOHBrOOBr

CAS RN 15086-94-9

IUPAC Name: 2',4',5',7'-tetrabromo-3',6'-dihydroxy-3H-spiro[2-benzofuran-1,9'-xanthen]-3-one
Synonyms: bromeosin solvent red 43 eosin acid eosin Y spirit soluble japan red 223 D and C red no. 21 bromoeosin japan red no. 223 C.I. solvent red 43 eosine acid
Molecular Weight (g/mol): 647.9
Molecular Formula: C20H8Br4O5
InChi Key: DBZJJPROPLPMSN-UHFFFAOYSA-N
SMILES: OC1=C(Br)C=C2C(OC3=C(Br)C(O)=C(Br)C=C3C22OC(=O)C3=CC=CC=C23)=C1Br

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results