CAS RN 1502-06-3
CAS RN 1502-06-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Cyclodecanone, TRC
CAS: 1502-06-3 Molecular Formula: C10H18O Molecular Weight (g/mol): 154.25 IUPAC Name: cyclodecanone SMILES: O=C1CCCCCCCCC1