CAS RN 14979-39-6
CAS RN 14979-39-6
IUPAC Name:
4-methylheptan-3-ol
Synonyms:
4-methylheptan-3-ol
Molecular Weight (g/mol):
130.23
Molecular Formula:
C8H18O
InChi Key:
BKQICAFAUMRYLZ-UHFFFAOYNA-N
SMILES:
CCCC(C)C(O)CC
1
–
1
of
1
results
1
4-Methyl-3-heptanol, erythro + threo, 99%
CAS: 14979-39-6 Molecular Formula: C8H18O Molecular Weight (g/mol): 130.231 MDL Number: MFCD00004568 InChI Key: BKQICAFAUMRYLZ-UHFFFAOYSA-N Synonym: 4-methyl-3-heptanol,3-heptanol, 4-methyl,caswell no. 570a,epa pesticide chemical code 571200,4-methyl-5-heptanol,acmc-1bqfj,dsstox_cid_21519,dsstox_rid_79763,dsstox_gsid_41519,3-heptanol, 4-methyl-van8c PubChem CID: 26989 IUPAC Name: 4-methylheptan-3-ol SMILES: CCCC(C)C(CC)O