CAS RN 147816-23-7
CAS RN 147816-23-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Cefcapene Pivoxil Hydrochloride, TRC
CAS: 147816-23-7 Molecular Formula: C23 H29 N5 O8 S2 . Cl H . H2 O Molecular Weight (g/mol): 622.11 IUPAC Name: 2,2-dimethylpropanoyloxymethyl (6R,7R)-7-[[(Z)-2-(2-amino-1,3-thiazol-4-yl)pent-2-enoyl]amino]-3-(carbamoyloxymethyl)-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate;hydrate;hydrochloride SMILES: O.Cl.CC\C=C(/C(=O)N[C@H]1[C@H]2SCC(=C(N2C1=O)C(=O)OCOC(=O)C(C)(C)C)COC(=O)N)\c3csc(N)n3