CAS RN 147657-22-5
CAS RN 147657-22-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
Calcipotriol Monohydrate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Calcipotriol Monohydrate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Calcipotriol monohydrate, MedChemExpress
MedChemExpress Calcipotriol monohydrate is a synthetic VitD3 analogue with a high affinity for the vitamin D receptor.