accessibility menu, dialog, popup

UserName

CAS RN 1415233-80-5

NHNSNH2Br

CAS RN 1415233-80-5

IUPAC Name: (6-bromopyridin-3-yl)thiourea
Synonyms: 6-bromopyridin-3-ylthiourea
Molecular Weight (g/mol): 232.1
Molecular Formula: C6H6BrN3S
InChi Key: SKISABIUKWHVJD-UHFFFAOYSA-N
SMILES: NC(=S)NC1=CN=C(Br)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results