CAS RN 13967-25-4
CAS RN 13967-25-4
IUPAC Name:
mercury(2+) difluoride
Synonyms:
mercury(II) fluoride mercury(I) fluoride
Molecular Weight (g/mol):
238.59
Molecular Formula:
F2Hg
InChi Key:
FMSYTQMJOCCCQS-UHFFFAOYSA-L
SMILES:
[F-].[F-].[Hg++]
1
–
1
of
1
results
1
Mercury(I) fluoride, 97+%, Thermo Scientific™
CAS: 13967-25-4 MDL Number: MFCD00016136