accessibility menu, dialog, popup

UserName

CAS RN 13958-98-0

NNBr

CAS RN 13958-98-0

IUPAC Name: 3-bromopyridine-4-carbonitrile
Synonyms: 3-bromopyridine-4-carbonitrile
Molecular Weight (g/mol): 183.01
Molecular Formula: C6H3BrN2
InChi Key: VPDIXJCBBUZNPO-UHFFFAOYSA-N
SMILES: BrC1=C(C=CN=C1)C#N
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)