CAS RN 13931-94-7
CAS RN 13931-94-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Chromium (II) Chloride Tetrahydrate (Technical Grade), TRC
CAS: 13931-94-7 Molecular Formula: CrCl2•4H2O Molecular Weight (g/mol): 194.96 Synonym: Chromium Chloride Tetrahydrate,Chromium Dichloride Tetrahydrate,Chromous Chloride Tetrahydrate IUPAC Name: chromium(II) chloride tetra hydrate SMILES: Cl[Cr]Cl.[4H2O]