CAS RN 1391054-52-6
CAS RN 1391054-52-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
7-Benzyloxy-2-(2,2-diphenyl-1,3-benzodioxol-5-yl)-5-hydroxy-H-1-benzopyran-4-one, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
7-Benzyloxy-2-(2,2-diphenyl-1,3-benzodioxol-5-yl)-5-hydroxy-H-1-benzopyran-4-one, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.