accessibility menu, dialog, popup

UserName

CAS RN 13709-52-9

FFHHHHFFHf

CAS RN 13709-52-9

IUPAC Name: tetrahydrogen hafnium tetrafluoride
Synonyms: hafnium tetrafluoride
Molecular Weight (g/mol): 258.52
Molecular Formula: F4H4Hf
InChi Key: HRTPXFIQXNXHQB-UHFFFAOYSA-N
SMILES: [H+].[H+].[H+].[H+].[F-].[F-].[F-].[F-].[Hf]
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (2)
  • (2)
  • (2)