CAS RN 1361644-26-9
CAS RN 1361644-26-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Aldoxorubicin, MedChemExpress
MedChemExpress Aldoxorubicin (INNO-206) is an albumin-binding prodrug of Doxorubicin (DNA topoisomerase II inhibitor), which is released from albumin under acidic conditions. Aldoxorubicin (INNO-206) has potent antitumor activities in various cancer cell lines and in murine tumor models.