accessibility menu, dialog, popup

UserName

CAS RN 13573-11-0

MgOWOOO

CAS RN 13573-11-0

IUPAC Name: magnesium(2+) dioxotungstenbis(olate)
Synonyms: magnesium(2+) tungstate
Molecular Weight (g/mol): 272.14
Molecular Formula: MgO4W
InChi Key: DJZHPOJZOWHJPP-UHFFFAOYSA-N
SMILES: [Mg++].[O-][W]([O-])(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results