accessibility menu, dialog, popup

UserName

CAS RN 13498-07-2

OClOOOOClOOOOClOOOPr

CAS RN 13498-07-2

IUPAC Name: praseodymium(3+) triperchlorate
Synonyms: praseodymium(3+) triperchlorate
Molecular Weight (g/mol): 439.25
Molecular Formula: Cl3O12Pr
InChi Key: OPAITELCMCAYEB-UHFFFAOYSA-K
SMILES: [Pr+3].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results