CAS RN 1329834-28-7
CAS RN 1329834-28-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Ibandronic Acid-d3 Sodium Salt, TRC
CAS: 1329834-28-7 Molecular Formula: C9 D3 H19 N O7 P2 . Na Molecular Weight (g/mol): 344.23 Synonym: [1-Hydroxy-3-(methyl-d3-pentylamino)propylidene]bisphosphnic Acid Sodium Salt IUPAC Name: sodium;hydroxy-[1-hydroxy-3-[pentyl(trideuteriomethyl)amino]-1-phosphono-propyl]phosphinate SMILES: [Na+].[2H]C([2H])([2H])N(CCCCC)CCC(O)(P(=O)(O)O)P(=O)(O)[O-]