accessibility menu, dialog, popup

UserName

CAS RN 1313-85-5

NaNaSe

CAS RN 1313-85-5

IUPAC Name: disodium selandiide
Synonyms: sodium selenide
Molecular Weight (g/mol): 124.95
Molecular Formula: Na2Se
InChi Key: VPQBLCVGUWPDHV-UHFFFAOYSA-N
SMILES: [Na+].[Na+].[Se--]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results