accessibility menu, dialog, popup

UserName

CAS RN 13091-23-1

ONONCl

CAS RN 13091-23-1

IUPAC Name: 4-chloro-3-nitropyridine
Synonyms: 4-chloro-3-nitropyridine
Molecular Weight (g/mol): 158.54
Molecular Formula: C5H3ClN2O2
InChi Key: JOTRPRKONYTVBV-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CN=CC=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results