accessibility menu, dialog, popup

UserName

CAS RN 129644-57-1

OFFFOHOOH

CAS RN 129644-57-1

IUPAC Name: 2-hydroxy-5-(trifluoromethoxy)benzoic acid
Synonyms: 2-hydroxy-5-(trifluoromethoxy)benzoic acid
Molecular Weight (g/mol): 222.12
Molecular Formula: C8H5F3O4
InChi Key: HNYMLXYADOZCNY-UHFFFAOYSA-N
SMILES: OC(=O)C1=C(O)C=CC(OC(F)(F)F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results