CAS RN 1294504-67-8
CAS RN 1294504-67-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
(R)-1-(5-Amino-2-(1-(benzyloxy)-2-methylpropan-2-yl)-6-fluoro-1H-indol-1-yl)-3-(benzyloxy)propan-2-ol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(R)-1-(5-Amino-2-(1-(benzyloxy)-2-methylpropan-2-yl)-6-fluoro-1H-indol-1-yl)-3-(benzyloxy)propan-2-ol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
(R)-1-(5-Amino-2-(1-(benzyloxy)-2-methylpropan-2-yl)-6-fluoro-1H-indol-1-yl)-3-(benzyloxy)propan-2-ol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.