CAS RN 128781-80-6
CAS RN 128781-80-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Boc-3,5-diiodo-tyr-ome, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.