accessibility menu, dialog, popup

UserName

CAS RN 127600-13-9

H3CH3COOO

CAS RN 127600-13-9

IUPAC Name: 4-(5,5-dimethyl-1,3-dioxan-2-yl)butanal
Synonyms: 4-(5,5-dimethyl-1,3-dioxan-2-yl)butanal
Molecular Weight (g/mol): 186.25
Molecular Formula: C10H18O3
InChi Key: KSKRSPYEXSAZRE-UHFFFAOYSA-N
SMILES: CC1(C)COC(CCCC=O)OC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results