CAS RN 126714-85-0
CAS RN 126714-85-0
IUPAC Name:
4,5-dichlorothiophene-2-sulfonyl chloride
Synonyms:
4,5-dichlorothiophene-2-sulfonyl chloride
Molecular Weight (g/mol):
251.52
Molecular Formula:
C4HCl3O2S2
InChi Key:
IVTWLTRKVRJPNG-UHFFFAOYSA-N
SMILES:
ClC1=C(Cl)C=C(S1)S(Cl)(=O)=O
1
–
1
of
1
results
1
2,3-Dichlorothiophene-5-sulfonyl chloride, 97%, Thermo Scientific™
CAS: 126714-85-0 Molecular Formula: C4HCl3O2S2 Molecular Weight (g/mol): 251.52 MDL Number: MFCD00052092 InChI Key: IVTWLTRKVRJPNG-UHFFFAOYSA-N Synonym: 2,3-dichlorothiophene-5-sulfonyl chloride,2,3-dichlorothiophene-5-sulphonyl chloride,4,5-dichloro-2-thiophenesulfonyl chloride,4,5-dichloro-thiophene-2-sulfonyl chloride,2-thiophenesulfonyl chloride, 4,5-dichloro,pubchem5523,acmc-20aoh3,4,5-dichloro 2-thienyl chlorosulfone,2,3-dichlorothiophene-5-sulfonylchloride,2-thiophenesulfonylchloride, 4,5-dichloro PubChem CID: 518622 IUPAC Name: 4,5-dichlorothiophene-2-sulfonyl chloride SMILES: ClC1=C(Cl)C=C(S1)S(Cl)(=O)=O