CAS RN 12627-53-1
CAS RN 12627-53-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
Thermo Scientific Chemicals Leishman Stain
CAS: 12627-53-1 MDL Number: MFCD00131498
2
Leishman's Stain MP Biomedicals
CAS: 12627-53-1 Synonym: Eosin-polychrome methylene blue