accessibility menu, dialog, popup

UserName

CAS RN 1262019-25-9

CH3OFFFOOH(E/Z)

CAS RN 1262019-25-9

IUPAC Name: 3-[2-methyl-5-(trifluoromethoxy)phenyl]prop-2-enoic acid
Synonyms: 3-[2-methyl-5-(trifluoromethoxy)phenyl]prop-2-enoic acid
Molecular Weight (g/mol): 246.19
Molecular Formula: C11H9F3O3
InChi Key: JVYBXYXIWXGYJQ-UHFFFAOYSA-N
SMILES: CC1=C(C=CC(O)=O)C=C(OC(F)(F)F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results