accessibility menu, dialog, popup

UserName

CAS RN 124-76-5

H3CH3C(S)(R)CH3(R)OH

CAS RN 124-76-5

IUPAC Name: (1R,2R,4S)-1,7,7-trimethylbicyclo[2.2.1]heptan-2-ol
Synonyms: isoborneol
Molecular Weight (g/mol): 154.25
Molecular Formula: C10H18O
InChi Key: DTGKSKDOIYIVQL-XKSSXDPKSA-N
SMILES: CC1(C)[C@H]2CC[C@@]1(C)[C@H](O)C2
Molecular Weight (g/mol) 
  • (6)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (3)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (3)
  • (3)
  • (6)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)