accessibility menu, dialog, popup

UserName

H3COCH3

CAS RN 123-19-3

IUPAC Name: heptan-4-one
Synonyms: propyl ketone 4-heptanone (natural) DI(N-propyl) ketone GBL
Molecular Weight (g/mol): 114.19
Molecular Formula: C7H14O
InChi Key: HCFAJYNVAYBARA-UHFFFAOYSA-N
SMILES: CCCC(=O)CCC
Molecular Weight (g/mol) 
  • (3)
  • (6)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (6)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (3)