accessibility menu, dialog, popup

UserName

CAS RN 122637-39-2

H3CONOOH

CAS RN 122637-39-2

IUPAC Name: 6-acetylpyridine-2-carboxylic acid
Synonyms: 6-acetylpyridine-2-carboxylic acid
Molecular Weight (g/mol): 165.15
Molecular Formula: C8H7NO3
InChi Key: SMFINMWLJMPOES-UHFFFAOYSA-N
SMILES: CC(=O)C1=NC(=CC=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results