CAS RN 12253-13-3
CAS RN 12253-13-3
IUPAC Name:
trialuminium manganese
Synonyms:
trialuminium manganese
Molecular Weight (g/mol):
135.88
Molecular Formula:
Al3Mn
InChi Key:
HNTUTSCEYJYKPS-UHFFFAOYSA-N
SMILES:
[Al].[Al].[Al].[Mn]
1
–
1
of
1
results
1
Manganese aluminide, 99.5% (metals basis), Thermo Scientific™
CAS: 12253-13-3 MDL Number: MFCD00213943 Synonym: aluminum, compd. with manganese 3:1,aluminum; manganese,trialumane manganese,aluminum, compound with manganese 3:1