accessibility menu, dialog, popup

UserName

CAS RN 121872-94-4

H3COClFF

CAS RN 121872-94-4

IUPAC Name: 1-(2-chloro-4,5-difluorophenyl)ethan-1-one
Synonyms: 1-(2-chloro-4,5-difluorophenyl)ethanone
Molecular Weight (g/mol): 190.57
Molecular Formula: C8H5ClF2O
InChi Key: BNODNMUCMFITCS-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC(F)=C(F)C=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results