CAS RN 12138-09-9
CAS RN 12138-09-9
IUPAC Name:
disulfanylidenetungsten
Synonyms:
tungsten disulfide tungsten(IV) sulfide tungsten disulphide
Molecular Weight (g/mol):
247.96
Molecular Formula:
S2W
InChi Key:
ITRNXVSDJBHYNJ-UHFFFAOYSA-N
SMILES:
S=[W]=S
1
–
1
of
1
results
1
Tungsten sulfide, 99.8% (metals basis)
CAS: 12138-09-9 Molecular Weight (g/mol): 247.96 MDL Number: MFCD00011467 InChI Key: ITRNXVSDJBHYNJ-UHFFFAOYSA-N IUPAC Name: disulfanylidenetungsten SMILES: S=[W]=S