accessibility menu, dialog, popup

UserName

CAS RN 120940-43-4

H3CNNH2NOOFFF

CAS RN 120940-43-4

IUPAC Name: 1-methyl-1-[2-nitro-4-(trifluoromethyl)phenyl]hydrazine
Synonyms: 1-methyl-1-[2-nitro-4-(trifluoromethyl)phenyl]hydrazine
Molecular Weight (g/mol): 235.17
Molecular Formula: C8H8F3N3O2
InChi Key: GBBAZCATUSUQOJ-UHFFFAOYSA-N
SMILES: CN(N)C1=CC=C(C=C1[N+]([O-])=O)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results