accessibility menu, dialog, popup

UserName

CAS RN 1206641-45-3

BOOH3CH3CCH3H3CNCH3OH

CAS RN 1206641-45-3

IUPAC Name: 2-[methyl({[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl})amino]ethan-1-ol
Synonyms: 2-[methyl({[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl})amino]ethanol
Molecular Weight (g/mol): 291.2
Molecular Formula: C16H26BNO3
InChi Key: PEMLGAZFFSJSCW-UHFFFAOYSA-N
SMILES: CN(CCO)CC1=CC=C(C=C1)B1OC(C)(C)C(C)(C)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results