accessibility menu, dialog, popup

UserName

CAS RN 12053-13-3

CrSe

CAS RN 12053-13-3

IUPAC Name: selanylidenechromium
Synonyms: chromium(II) selenide
Molecular Weight (g/mol): 130.97
Molecular Formula: CrSe
InChi Key: UVZCKRKEVWSRGT-UHFFFAOYSA-N
SMILES: [Cr]=[Se]
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results