accessibility menu, dialog, popup

UserName

CAS RN 12032-74-5

OOOTiMn

CAS RN 12032-74-5

IUPAC Name: titanium(4+) manganese(2+) trioxidandiide
Synonyms: manganese(II) titanate
Molecular Weight (g/mol): 150.8
Molecular Formula: MnO3Ti
InChi Key: IXZOTKANSDQAHZ-UHFFFAOYSA-N
SMILES: [O--].[O--].[O--].[Ti+4].[Mn++]
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results