CAS RN 12022-99-0
CAS RN 12022-99-0
IUPAC Name:
(silanylideneferrio)silane
Synonyms:
(silanylideneferrio)silane
Molecular Weight (g/mol):
116.05
Molecular Formula:
FeH4Si2
InChi Key:
UVUDQHYBXBVHAI-UHFFFAOYSA-N
SMILES:
[SiH2]=[Fe]=[SiH2]