CAS RN 1196-31-2
CAS RN 1196-31-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(+)-Isomenthone, TRC
CAS: 1196-31-2 Molecular Formula: C10 H18 O Molecular Weight (g/mol): 154.25 Synonym: (+)-Isomenthone,(2R,5R)-2-Isopropyl-5-methyl-cyclohexanone,(1R,4R)-Isomenthone,(1R,4R)-p-Menth-3-one,D-Isomenthone,L-Isomenthone,d-Isomenthone,d-Menthone IUPAC Name: (2R,5R)-5-methyl-2-propan-2-ylcyclohexan-1-one SMILES: CC(C)[C@H]1CC[C@@H](C)CC1=O