accessibility menu, dialog, popup

UserName

CAS RN 1187931-09-4

NNONH2HCl

CAS RN 1187931-09-4

IUPAC Name: 1-(5-phenyl-1,3,4-oxadiazol-2-yl)methanamine hydrochloride
Synonyms: 1-(5-phenyl-1,3,4-oxadiazol-2-yl)methanamine hydrochloride
Molecular Weight (g/mol): 211.65
Molecular Formula: C9H10ClN3O
InChi Key: MFFARWJMWVFLIW-UHFFFAOYSA-N
SMILES: Cl.NCC1=NN=C(O1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results