accessibility menu, dialog, popup

UserName

CAS RN 117924-33-1

H3CNH3CPOCH3H3CCH3OCH3H3CCH3

CAS RN 117924-33-1

IUPAC Name: [bis(tert-butoxy)phosphanyl]diethylamine
Synonyms: [bis(tert-butoxy)phosphanyl]diethylamine
Molecular Weight (g/mol): 249.34
Molecular Formula: C12H28NO2P
InChi Key: KUKSUQKELVOKBH-UHFFFAOYSA-N
SMILES: CCN(CC)P(OC(C)(C)C)OC(C)(C)C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)