CAS RN 1165-57-7
CAS RN 1165-57-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
o,m-Quaterphenyl, TRC
CAS: 1165-57-7 Molecular Formula: C24 H18 Molecular Weight (g/mol): 306.4 Synonym: 2,3'-Diphenylbiphenyl,1,1':2',1' ':3' ',1' ' '-Quaterphenyl IUPAC Name: 1-phenyl-2-(3-phenylphenyl)benzene SMILES: c1ccc(cc1)c2cccc(c2)c3ccccc3c4ccccc4