CAS RN 116490-30-3
CAS RN 116490-30-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
4-Aminobiphenyl beta-D-Glucuronide Sodium Salt (>85%), TRC
CAS: 116490-30-3 Molecular Formula: C18 H18 N Na O6 Molecular Weight (g/mol): 367.33 Synonym: 1-([1,1'-Biphenyl]-4-ylamino)-1-deoxy-β-D-glucopyranuronic Acid Sodium Salt; IUPAC Name: sodium;(2S,3S,4S,5R,6R)-3,4,5-trihydroxy-6-(4-phenylanilino)tetrahydropyran-2-carboxylate SMILES: [Na+].O[C@H]1[C@H](Nc2ccc(cc2)c3ccccc3)O[C@@H]([C@@H](O)[C@@H]1O)C(=O)[O-]