accessibility menu, dialog, popup

UserName

CAS RN 115687-05-3

NNNNOOOCl

CAS RN 115687-05-3

IUPAC Name: 5-[4-(4-chlorobenzoyl)piperazin-1-yl]-8-nitroquinoline
Synonyms: 5-[4-(4-chlorobenzoyl)piperazin-1-yl]-8-nitroquinoline
Molecular Weight (g/mol): 396.83
Molecular Formula: C20H17ClN4O3
InChi Key: KLNPVNZJCWIQSK-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(N2CCN(CC2)C(=O)C2=CC=C(Cl)C=C2)C2=CC=CN=C12
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results